missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alclofenac Methyl Ester, Mikromol™
Description
Alclofenac Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Alclofenac Methyl Ester |
| CAS | 23918-07-2 |
| Analyte or Component Names | Alclofenac Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H13 Cl O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Alclofenac Methyl Ester, 3-Chloro-4-(2-propen-1-yloxy)benzeneacetic acid methyl ester, Acetic acid, [4-(allyloxy)-3-chlorophenyl]-, methyl ester (8CI), Benzeneacetic acid, 3-chloro-4-(2-propenyloxy)-, methyl ester (9CI), Alclofenac methyl ester |
| InChI Formula | InChI=1S/C12H13ClO3/c1-3-6-16-11-5-4-9(7-10(11)13)8-12(14)15-2/h3-5,7H,1,6,8H2,2H3 |
| SMILES | COC(=O)Cc1ccc(OCC=C)c(Cl)c1 |
| IUPAC Name | methyl 2-(3-chloro-4-prop-2-enoxyphenyl)acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?