Learn More
Alclofenac, LoGiCal
Product Code. 30461975
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461975 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461975 Supplier LoGiCal Supplier No. LGCFOR1783.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Alclofenac
CAS 22131-79-9
Analyte or Component Names Alclofenac
Shipping Condition Room Temperature
Molecular Formula C11 H11 Cl O3
Certifications/Compliance ISO 9001
Synonym Benzeneacetic acid, 3-chloro-4-(2-propen-1-yloxy)-Acetic acid, [4-(allyloxy)-3-chlorophenyl]- (8CI), Benzeneacetic acid, 3-chloro-4-(2-propenyloxy)- (9CI), Aclofenac, Alclofenac, Alclophenac, Allopydin, Epinal, MY 101, Medifenac, Mervan, Neosten, Neoston, Prinalgin, Reufenac, W 7320, Zumaril, [3-Chloro-4-(allyloxy)phenyl]acetic acid, [4-(Allyloxy)-3-chlorophenyl]acetic acid
InChI Formula InChI=1S/C11H11ClO3/c1-2-5-15-10-4-3-8(6-9(10)12)7-11(13)14/h2-4,6H,1,5,7H2,(H,13,14)
Solution Type Neat
SMILES OC(=O)Cc1ccc(OCC=C)c(Cl)c1
IUPAC Name 2-(3-chloro-4-prop-2-enoxyphenyl)acetic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 226.66
Quantity 10 mg
Formula Weight 226.0397 g/mol