Learn More
Alclofenac, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Alclofenac |
| CAS | 22131-79-9 |
| Analyte or Component Names | Alclofenac |
| Shipping Condition | +20°C |
| Molecular Formula | C11 H11 Cl O3 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Benzeneacetic acid, 3-chloro-4-(2-propen-1-yloxy)-Acetic acid, [4-(allyloxy)-3-chlorophenyl]- (8CI), Benzeneacetic acid, 3-chloro-4-(2-propenyloxy)- (9CI), Aclofenac, Alclofenac, Alclophenac, Allopydin, Epinal, MY 101, Medifenac, Mervan, Neosten, Neoston, Prinalgin, Reufenac, W 7320, Zumaril, [3-Chloro-4-(allyloxy)phenyl]acetic acid, [4-(Allyloxy)-3-chlorophenyl]acetic acid |
| InChI Formula | InChI=1S/C11H11ClO3/c1-2-5-15-10-4-3-8(6-9(10)12)7-11(13)14/h2-4,6H,1,5,7H2,(H,13,14) |
| SMILES | OC(=O)Cc1ccc(OCC=C)c(Cl)c1 |
| IUPAC Name | 2-(3-chloro-4-prop-2-enoxyphenyl)acetic acid |
| Show More |