missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ALCAM/CD166 Antibody (praluzatamab) - Humanized, Novus Biologicals™

Human Monoclonal Antibody

213.00€ - 920.00€

Specifications

Antigen ALCAM/CD166
Clone praluzatamab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30045852
View Documents
Novus Biologicals™
NBP3-28283-1mg
1 mg
920.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044297
View Documents
Novus Biologicals™
NBP3-28283-50ug
50 μg
213.00€
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30047247
View Documents
Novus Biologicals™
NBP3-28283-100ug
100 μg
326.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

ALCAM/CD166
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
ALCAM/ CD166
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
praluzatamab
Monoclonal
Purified
RUO
Human
Q13740
214
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.