missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALCAM/CD166 Antibody (5B3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22211-0.025ml
This item is not returnable.
View return policy
Description
ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ALCAM/CD166 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| ALCAM | |
| Purified recombinant fragment of human CD166 (AA: 405-524) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5B3 | |
| Unconjugated | |
| activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
| Mouse | |
| 65.1 kDa | |
| 0.025 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 214 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction