missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Albumin Native Protein
Shop All Bio Techne Products
Click to view available options
:
1mg; Rhodamine
Description
Native Albumin. Source: Serum The Albumin Native Protein has been validated for the following applications: Western Blot, Dot Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.
Specifications
Specifications
| For Use With (Application) | Dot Blot |
| Formulation | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Polyethylene Glycol (PEG-8000) |
| Gene ID (Entrez) | 213 |
| Name | Albumin protein |
| Storage Requirements | 4°C in the dark. |
| Cross Reactivity | Human |
| Purity or Quality Grade | Chromatography |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction