Learn More
MP Biomedicals™ Albumin, Bovine, For IVD, Low Ash and Metals
Product Code. 11420262 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
50g
Product Code. 11420262
Brand: MP Biomedicals™ 0216006990
Prepared by heat shock fractionation
  • For IVD applications
  • Low visible absorbance
  • Purity: >98%
  • Ash: <2%
  • Metals: <20ppm
For best cell culture performance, see alternate Mfr. No. 180577
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) ELISA, Microbial Culture
Molecular Weight (g/mol) 66.338
Name Bovine Albumin Fraction V
pH Range 6.5 to 7.5
Quantity 500 g
Source Bovine
Storage Requirements 2° to 8°C
Form Powder