Learn More
MP Biomedicals™ Albumin, Bovine, For IVD, Low Ash and Metals
Shop All MP Biomedical Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
50g
Description
- For IVD applications
- Low visible absorbance
- Purity: >98%
- Ash: <2%
- Metals: <20ppm
For best cell culture performance, see alternate Mfr. No. 180577
Specifications
Specifications
| For Use With (Application) | ELISA, Microbial Culture |
| Molecular Weight (g/mol) | 66.338 |
| Name | Bovine Albumin Fraction V |
| pH Range | 6.5 to 7.5 |
| Quantity | 100 g |
| Source | Bovine |
| Storage Requirements | 2° to 8°C |
| Form | Powder |