missing translation for 'onlineSavingsMsg'
Learn More

Albumin Antibody - BSA Free, Novus Biologicals™

Product Code. 18235010 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18235010 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18235010 Supplier Novus Biologicals Supplier No. NB300336

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

Albumin Polyclonal specifically detects Albumin in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, ELISA, Aggregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Albumin
Applications Western Blot, ELISA, Aggregation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Aggregation
Formulation PBS with 0.02% Sodium Azide
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Chicken
Immunogen Recombinant serum albumin (GI # 113576) sequence 1-609
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 213
Test Specificity Albumin
Target Species Human, Mouse, Rat, Pig, Bovine, Canine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.