missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1, p Ser473 Antibody (104A282) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB100-56749
This item is not returnable.
View return policy
Description
AKT1 Monoclonal specifically detects AKT1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| AKT1 (p Ser473) | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry 1:200 - 1:250, Immunohistochemistry-Paraffin 1:200 - 1:250, Immunohistochemistry-Frozen reported by customer review | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
| Mouse | |
| 55.7 kDa | |
| 0.1 mg | |
| Angiogenesis, Apoptosis, Autophagy, Cancer, Cellular Markers, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Serine Kinases, Signal Transduction, Tyrosine Kinases | |
| 207 | |
| Human, Mouse, Rat, Rabbit | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 104A282 | |
| Unconjugated | |
| P31749 | |
| AKT1 | |
| This AKT1 phospho Ser473 antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 473 of human AKT1. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Clone 104A282 detects specifically the Ser473 phosphorylated form of AKT1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction