Learn More
Syngene™ AirClean™ Microscope Enclosure with UVTect™ Microprocessor Controller
Shop All Syngene Products
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
Description
Standard Features
- No ductwork required
- No installation required - plugs directly into a standard 110V or 220V electrical outlet
- Encloses equipment while retaining full operator access
- UVTect™ Microprocessor Controller
- ISO 5 vertical laminar flow air
- UV and fluorescent lights
- Seamless plastic design - no joints or gaps in construction
- Lab event timer
- HEPA filtration for capture of particulate to 0.3 microns in size
- Filter safety lockout to ensure correct filter replacement
- Polycarbonate and polypropylene design reflects UV energy
- Tested to UL 61010-1,CSA C22.2 No. 61010-1, and EN 61326-1
- Brushless, sparkless blower is post-filter
- Filter and electronics are located above the workspace, eliminating the chance of chemical spill
- Real time monitoring of installed filtration bed
Specifications
Specifications
| Product Type | Microscope Enclosure |
| Includes | UVTect™ Microprocessor Controller |
| Width (English) | 48 in. |
| Width (Metric) | 1220 mm |