missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIPL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38272-20ul
This item is not returnable.
View return policy
Description
AIPL1 Polyclonal antibody specifically detects AIPL1 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| AIPL1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| AIPL2, aryl hydrocarbon receptor interacting protein-like 1, aryl hydrocarbon receptor-interacting protein-like 1, aryl-hydrocarbon-interacting protein-like 1, LCA4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-384 of human AIPL1 (NP_055151.3).,, Sequence:, MDAALLLNVEGVKKTILHGGTGELPNFITGSRVIFHFRTMKCDEERTVIDDSRQVGQPMHIIIGNMFKLEVWEILLTSMRVHEVAEFWCDTIHTGVYPILSRSLRQMAQGKDPTEWHVHTCGLANMFAYHTLGYEDLDELQKEPQPLVFVIELLQVDAPSDYQRETWNLSNHEKMKAVPVLHGEGNRLFKLGRYEEASSKYQEAIICLRNLQTKEKPWEVQWLKLEKMINTLILNYCQCLLKKEEYYEVLEHTSDILRHHPGIVKAYYVRARAHAEVWNEAEAKADLQKVLELEPSMQKAVRRELRLLENRMAEKQEEERLRCRNMLSQGATQPPAEPPTEPPAQSSTEPPAEPPTAPSAELSAGPPAEPATEPPPSPGHSLQH | |
| 20 μL | |
| Vision | |
| 23746 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction