missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AIF-1/Iba1 Polyclonal specifically detects AIF-1/Iba1 in Human, Mouse, Rat, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockout Validated, Dual RNAscope ISH-IHC.
Specifications
Specifications
| Antigen | AIF-1/Iba1 |
| Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Dilution | Western Blot 0.1 - 1.0 ug/mL, Immunohistochemistry 4 - 6 ug/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 4 - 6 ug/mL, Immunohistochemistry-Frozen, Peptide ELISA Detection Limit 1:128000, Immunohistochemistry Free-Floating, Immunohistochemistry (Whole-Mount), Knockout Validated, Dual RNAscope ISH-IHC |
| Purification Method | Affinity Purified |
| Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?