missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AIDA
GREENER_CHOICE

Product Code. p-4783237
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18085172

Brand: Tocris Bioscience™ 0904/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective group I mGlu antagonist

A relatively potent and selective antagonist of group I metabotropic glutamate receptors (mGlu1a), having no effect on group II (mGlu2) or group III (mGlu4) receptors expressed individually in baby hamster kidney cells. Has no effect on ionotropic glutamate receptors. Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168560-79-0
Molecular Formula C11H10NO4

Specifications

Quantity 10 mg
Formula Weight 221.21
Purity >98%
Molecular Weight (g/mol) 220.21
Chemical Name or Material AIDA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.