accessibility menu, dialog, popup

UserName

CAS RN 168560-79-0

NH3(R)OOOO

CAS RN 168560-79-0

IUPAC Name: (1R)-1-azaniumyl-2,3-dihydro-1H-indene-1,5-dicarboxylate
Synonyms: (1R)-1-ammonio-2,3-dihydroindene-1,5-dicarboxylate
Molecular Weight (g/mol): 220.21
Molecular Formula: C11H10NO4
InChi Key: LSTPKMWNRWCNLS-LLVKDONJSA-M
SMILES: [NH3+][C@@]1(CCC2=CC(=CC=C12)C([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)