missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00€ - 430.00€
Specifications
| Antigen | AHR |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18420898
|
Novus Biologicals
NBP1-89974-25ul |
25 μL |
302.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18498365
|
Novus Biologicals
NBP1-89974 |
0.1 mL |
430.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AHR Polyclonal antibody specifically detects AHR in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| AHR | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Chromatin Research, Hypoxia, Innate Immunity, Transcription Factors and Regulators | |
| PBS (pH 7.2) and 40% Glycerol | |
| 196 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NWQDNTAPMGNDTILKHEQIDQPQDVNSFAGGHPGLFQDSKNSDLYSIMKNLGIDFEDIRHMQNEKFFRNDFSGEVDFRDIDLTDEI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title