Learn More
AGMAT Antibody, Novus Biologicals™
Product Code. 18730724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18730724 0.1 mL 0.1mL
18424521 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18730724 Supplier Novus Biologicals Supplier No. NBP182080
Rabbit Polyclonal Antibody
AGMAT Polyclonal specifically detects AGMAT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:MLGTVNPSTGALPFQSLMVADLGDVNVNLYNLQDSCRRIQEAYEKIVAAGCIPLTLGGDHTITYPILQAMAKKHGPVGLLHVDAHTDTTDKALGEKLYHG
Quantity 0.1 mL