Learn More
AGFG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12670-1-AP-20UL
 View more versions of this product
Product Code. 16850514
  • 145.00€ / 20µL
Description
The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintece of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2,and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
Specifications
Specifications
AGFG1
Polyclonal
Unconjugated
AGFG1
AGFG1, ArfGAP with FG repeats 1, HIV 1 Rev binding protein, HRB, Nucleoporin like protein RIP, RAB, Rev interacting protein, RIP
Rabbit
Antigen Affinity Chromatography
RUO
15463, 3267, 363266
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52594, Q4KLH5, Q8K2K6
AGFG1
AGFG1 Fusion Protein Ag3352
20 μL
Primary
Mouse, Human, Rat
Antibody