missing translation for 'onlineSavingsMsg'
Learn More

AGAP1 Antibody (OTI4E8), CoraFluor™ 1, Novus Biologicals™

Product Code. 30147318
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30147318 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30147318 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP272357CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AGAP1 Monoclonal antibody specifically detects AGAP1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGAP1
Applications Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
Classification Monoclonal
Clone OTI4E8
Conjugate CoraFluor 1
Formulation PBS
Gene Alias AGAP-1, ANK repeat and PH domains 1, ArfGAP with GTPase domain, ankyrin repeat and PH domain 1, centaurin, gamma 2, centaurin-gamma-2, CENTG2, cnt-g2, GTP-binding and GTPase-activating protein 1, KIAA1099arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1, MGC71657
Host Species Mouse
Immunogen Full length human recombinant protein of human AGAP1(NP_055729) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116987
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.