missing translation for 'onlineSavingsMsg'
Learn More

Abcam Ag 825, ErbB2 Inhibitor, Solid, 10 mg

Product Code. 32120820
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120820 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120820 Supplier Abcam Supplier No. ab14556610mg
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective ErbB2 inhibitor that suppresses IL-6 induced STAT3 activation and exhibits antitumor effects in vivo.

  • Molecular Weight: 397.5 Da
  • Purity: >96%
  • Form: Solid
  • Activity: Selective ErbB2 inhibitor (IC50 values are 0.15 μM at ErbB2 and 19 μM at ErbB1)
  • Biological effects: Suppresses IL-6 induced STAT3 activation, shows antiproliferative effects, and shows antitumor effects in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Alternate Part Number 149092-50-2
Buffer 100% AG 825
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule ErbB2, ErbB1
Formulation 100% AG 825
Quantity 10 mg
For Use With (Application) ErbB2 inhibition, STAT3 activation suppression, antiproliferative research, antitumor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.