Learn More
AffiniPure Rabbit anti-Goat, Biotin, Polyclonal, Proteintech
Product Code. 17224916 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17224916 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17224916 Supplier Proteintech Supplier No. SA000044200UL
 Restricted Item
In Stock
Add to basket
Add to basket
Rabbit Polyclonal Antibody
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
Antigen AffiniPure
Applications ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Biotin
Host Species Rabbit
Quantity 200 μL
Primary or Secondary Secondary
Target Species Human, Mouse, Rat
Form Liquid
Isotype IgG