Learn More
AffiniPure Goat anti-Rabbit, HRP, Polyclonal, Proteintech
Product Code. 17274896 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17274896 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17274896 Supplier Proteintech Supplier No. SA000012200UL
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Goat Polyclonal Antibody
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse samples. It is validated for ELISA, Western Blot.
Specifications
Antigen AffiniPure
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate HRP
Host Species Goat
Quantity 200 μL
Primary or Secondary Secondary
Target Species Human, Mouse
Form Liquid
Isotype IgG