Learn More
AffiniPure Goat anti-Mouse, AP, Polyclonal, Proteintech
Product Code. 17214906 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17214906 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17214906 Supplier Proteintech Supplier No. SA000021200UL
Goat Polyclonal Antibody
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse samples. It is validated for ELISA, Western Blot.
Specifications
Antigen AffiniPure
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Alkaline Phosphatase (AP)
Host Species Goat
Quantity 200 μL
Primary or Secondary Secondary
Target Species Human, Mouse
Form Liquid
Isotype IgG