missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP1L2 Antibody (4E12D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61837-0.025ml
This item is not returnable.
View return policy
Description
AFAP1L2 Monoclonal specifically detects AFAP1L2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| AFAP1L2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4 | |
| Mouse | |
| 91.3 kDa | |
| 0.025 mL | |
| 84632 | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 4E12D7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AFAP1L2 | |
| Purified recombinant fragment of human AFAP1L2 (AA: 674-818) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction