missing translation for 'onlineSavingsMsg'
Learn More

AFAP1L2 Antibody (2H6D1) - BSA Free, Novus Biologicals™

Product Code. 18696768 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18696768 0.025 mL 0.03mL
18279661 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18696768 Supplier Novus Biologicals Supplier No. NBP2618360.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AFAP1L2 Monoclonal specifically detects AFAP1L2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFAP1L2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2H6D1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4
Gene Symbols AFAP1L2
Host Species Mouse
Immunogen Purified recombinant fragment of human AFAP1L2 (AA: 674-818) expressed in E. Coli.
Molecular Weight of Antigen 89.8 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Primary or Secondary Primary
Gene ID (Entrez) 84632
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.