missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3B4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35765-20ul
This item is not returnable.
View return policy
Description
SF3B4 Polyclonal antibody specifically detects SF3B4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SF3B4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human SF3B4 (NP_005841.1).,, Sequence:, MAAGPISERNQDATVYVGGLDEKVSEPLLWELFLQAGPVVNTHMPKDRVTGQHQGYGFVEFLSEEDADYAIKIMNMIKLYGKPIRVNKASAHNKNLDVGANIFIGNLDPEIDEKLLYDTFSAFGVILQTPKIMRDPDTGNSKGYAFINFASFDASDAAIEAMNGQYLCNRPITVSYAFKKDSKGERHGSAAERLLAAQNP | |
| 20 μL | |
| Neuroscience | |
| 10262 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction