Learn More
Eppendorf™ Aerosol-Tight Rotor Lids
Product Code. 10386842 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
FA–45-24-11
FA–45-30-11
Rotor A-4-81
Unit Size:
Each
Pack of 2
Product Code. 10386842
Brand: Eppendorf™ 5490025806
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Providing secure closure for a variety of Eppendorf rotors.
  • Aluminim construction
  • Aerosol-tight
  • Autoclavable for 20 min at 121°C
TRUSTED_SUSTAINABILITY
Specifications
Type Aerosol-tight Lid
Material Aluminum
For Use With (Equipment) FA–45-30-11
Autoclavable Autoclavable (121°C for 20 min.)