Learn More
Eppendorf™ Aerosol-Tight Caps
Use these replacement lids with a variety of Eppendorf™ buckets.
Brand:  Eppendorf™ 5804712005
115.54 EUR valid until 2023-12-31
Use promo code "20333" to get your promotional price.
Additional Details : Weight : 0.17100kg
 View more versions of this product
Product Code. 10063201
  • 127.00€ / Pack of 2
Estimated Shipment: 26-06-2023
Description
  • Set of 2 caps 
  • Aerosol tight
Specifications
Specifications
Centrifuge Tube Cap
100mL Rectangular Bucket