missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00077PE
This item is not returnable.
View return policy
Description
ADK Polyclonal antibody specifically detects ADK in Human samples. It is validated for Western Blot, ELISA
Specifications
| ADK | |
| Polyclonal | |
| Western Blot, ELISA | |
| adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
| Produced in rabbits immunized with purified, recombinant Human ADK (Accession#: AAH03568.1; Met 1-His 345). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism | |
| 132 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction