missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AdipoR1 Antibody (2416C) [PE], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10128P
This item is not returnable.
View return policy
Description
AdipoR1 Monoclonal antibody specifically detects AdipoR1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| AdipoR1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 2416C | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| ACDCR1, adiponectin receptor 1, adiponectin receptor protein 1, ADR1, CGI45, FLJ42464, PAQR1FLJ25385, Progestin and adipoQ receptor family member I, TESBP1A | |
| Synthetic peptide containing human AdipoR1 C-term sequences, | |
| 0.1 mL | |
| Signal Transduction | |
| 51094 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction