missing translation for 'onlineSavingsMsg'
Learn More

Adiponectin/Acrp30 Antibody (S03-9F3), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00€ - 645.00€

Specifications

Antigen Adiponectin/Acrp30
Clone S03-9F3
Concentration 0.3 mg/mL
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18681715
View Documents
Novus Biologicals
NBP3-20028-100ul
100 μg
645.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18633074
View Documents
Novus Biologicals
NBP3-20028-50ul
50 μg
421.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Adiponectin/Acrp30 Monoclonal antibody specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications

Specifications

Adiponectin/Acrp30
0.3 mg/mL
Monoclonal
Purified
RUO
Human
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
A synthetic peptide of human Adiponectin/Acrp30 (Uniprot # Q15848)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S03-9F3
Western Blot, ELISA, Sandwich ELISA
Unconjugated
Rabbit
Diabetes Research, Lipid and Metabolism
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
9370
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.