missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylosuccinate Synthase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 462.00€
Specifications
| Antigen | Adenylosuccinate Synthase |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Adenylosuccinate Synthase Polyclonal antibody specifically detects Adenylosuccinate Synthase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Adenylosuccinate Synthase | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 159 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ADEH, adenylosuccinate synthase, adenylosuccinate synthetase (Ade(-)H-complementing), adenylosuccinate synthetase isozyme 2, Adenylosuccinate synthetase, acidic isozyme, Adenylosuccinate synthetase, liver isozyme, AdSS 2, ADSS2, AMPSase 2, EC 6.3.4.4, IMP--aspartate ligase 2, L-type adenylosuccinate synthetase, MGC20404 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ADSS (NP_001117.2). MAFAETYPAASSLPNGDCGRPRARPGGNRVTVVLGAQWGDEGKGKVVDLLAQDADIVCRCQGGNNAGHTVVVDSVEYDFHLLPSGIINPNVTAFIGNGVVIHLPGLFEEAEKNVQKGKGLEGWEKRLIISDRAHIVFDFHQAADGIQEQQRQEQAGKNLGTTKKGIGPVYSSKAARSGLRMCDLVSDFDGFSERFKVLANQYKSIYPTLEIDIEGELQKLKGYMEKIKPMVRDGVYFLYEALHGPPKKILVEGANAALLDIDFGTYPFVTSSNCTVGGVC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title