missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAR Antibody (CL0176) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05500MFV500
This item is not returnable.
View return policy
Description
ADAR Monoclonal antibody specifically detects ADAR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| ADAR | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| ADAR1DSH, adenosine deaminase, RNA-specific, DRADAP136, DSRADadenosine deaminase acting on RNA 1-A, EC 3.5.4, EC 3.5.4.-, G1P1double-stranded RNA-specific adenosine deaminase, IFI-4, IFI4dsRNA adenosine deaminase, interferon-induced protein 4, Interferon-inducible protein 4, K88DSRBP, p136,136 kDa double-stranded RNA-binding protein | |
| This antibody was developed using a recombinant protein derived from P55265, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0176 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 103 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction