Learn More
Eppendorf™ Adapters For Fixed-Angle Round Bottom Rotor
Product Code. 10450332 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
15 to 18 mL
5 mL
7 to 15 mL
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10450332 5 mL Pack of 2
10053311 7 to 15 mL Pack of 2
10249351 15 to 18 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10450332 Supplier Eppendorf™ Supplier No. 5804774000
Estimated Shipment: 07-12-2026
Add flexibility to Eppendorf Centrifuges with these rotor adapters
  • Adapters
  • For Eppendorf™ Fixed Angle Round-Bottom Rotors
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Round Bottom
Capacity 5 mL
For Use With (Equipment) Fixed angle rotor
Bore Size (Metric) 29 mm
No. of Places 6
No. per Pack 2 Pack