Learn More
Eppendorf™ Adapters for Rectangular Buckets in A-4-81 Rotor
Product Code. p-8003413 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
15 mL
180 to 250 mL
2.6 to 7 mL
3 to 15 mL
Unit Size:
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11830242 15 mL Pack of 2
11833981 180 to 250 mL Pack of 2
11803432 3 to 15 mL Pack of 2
10137533 2.6 to 7 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11830242 Supplier Eppendorf™ Supplier No. 5810722004
Only null left
Add to basket
Add to basket
Add flexibility to A-4-81 Rotor for Eppendorf Centrifuge 5810/5810 R with these adapters
  • Rotor adapters
  • For rectangluar buckets
  • Available in a wide range of sizes to suit various applications
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Conical
Capacity 15 mL
For Use With (Equipment) 400mL Rectangular Bucket for Rotor A-4-81
Bore Size (Metric) 17.5 mm
No. of Places 48
No. per Pack 2 Pack