Learn More
Eppendorf™ Adapters For 100mL Rectangular Bucket
Product Code. 10618631 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1.2 to 5 mL
1.5 to 2 mL
18 to 120 mL
18 to 30 mL
2.6 to 7 mL
3 to 15 mL
30 to 50 mL
50 to 75 mL
7 to 18 mL
Unit Size:
Pack of 2
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10618631 2.6 to 7 mL Pack of 2
10266861 1.2 to 5 mL Pack of 2
10555921 1.5 to 2 mL Pack of 2
10318611 50 to 75 mL Pack of 2
10681502 18 to 120 mL Pack of 2
10786722 30 to 50 mL Pack of 2
10648821 18 to 30 mL Pack of 2
10297581 7 to 18 mL Pack of 2
10247251 3 to 15 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10618631 Supplier Eppendorf™ Supplier No. 5804752007
Estimated Shipment: 07-12-2026
For use with Eppendorf Tubes
  • Adapters for rectangular buckets
  • For use with Rotor A-4-44
  • Available in a variety of sizes for a wide range of applications
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Capacity 2.6 to 7 mL
For Use With (Equipment) 100mL Rectangular Bucket
Bore Size (Metric) 13 mm
No. of Places 36
For Use With (Application) Standard, Vacutainer, Falcon and micro test tubes
No. per Pack 2 Pack