Learn More
Macherey-Nagel™ Adapters for Chromabond™ SPE Columns
Product Code. 12471498
Click to view available options
For Use With:
Glass columns (3 and 6mL)
PP columns (15, 45 and 70mL)
PP columns (30 and 70mL)
Material:
PVDF
Polyethylene
Unit Size:
Each
Product Code. 12471498
Brand: Macherey-Nagel™ 730566
Only null left
Add to basket
Add to basket
For individual packing of SPE columns with Chromabond™ adsorbents. Macherey-Nagel™ Adapters for Chromabond™ SPE Columns are available in several materials for a variety of empty column types and capacities.
  • PVDF adapter for Glass columns (3 and 6mL)
  • PE adapter for Polypropylene columns (15, 45 and 70mL)
  • PE adapter for Polypropylene columns (30 and 70mL)
Specifications
Type Adapter
For Use With (Application) Solid Phase Extraction (SPE)
For Use With PP columns (30 and 70mL)
Material Polyethylene