Learn More
Macherey-Nagel™ Adapters for Chromabond™ SPE Columns
Product Code. 12446546
Click to view available options
For Use With:
Glass columns (3 and 6mL)
PP columns (15, 45 and 70mL)
PP columns (30 and 70mL)
Material:
PVDF
Polyethylene
Unit Size:
Each
Product Code. 12446546
Brand: Macherey-Nagel™ 730104.4
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For individual packing of SPE columns with Chromabond™ adsorbents. Macherey-Nagel™ Adapters for Chromabond™ SPE Columns are available in several materials for a variety of empty column types and capacities.
  • PVDF adapter for Glass columns (3 and 6mL)
  • PE adapter for Polypropylene columns (15, 45 and 70mL)
  • PE adapter for Polypropylene columns (30 and 70mL)
Specifications
Type Tubing Adapter
For Use With (Application) Solid Phase Extraction (SPE)
For Use With Glass columns (3 and 6mL)
Material PVDF