missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM10 Antibody (CL13808) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24577MFV500
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal antibody specifically detects ADAM10 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ADAM10 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| This antibody was developed using a synthetic peptide derived from O14672, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Signal Transduction | |
| 102 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL13808 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction