missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ACVR1B (Human) Recombinant Protein

Human ACVR1B full-length ORF ( AAH00254.1, 1 a.a. - 505 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000091-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101241

  • 335.00€ / 10µg

Estimated Shipment: 11-05-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MAESAGASSFFPLVVLLLAGSGGSGPRGVQALLCACTSCLQANYTCETDGACMVSIFNLDGMEHHVRTCIPKVELVPAGKPFYCLSSEDLRNTHCCYTDYCNRIDLRVPSGHLKEPEHPSMWGPVELVGIIAGPVFLLFLIIIIVFLVINYHQRVYHNRQRLDMEDPSCEMCLSKDKTLQDLVYDLSTSGSGSGLPLFVQRTVARTIVLQEIIGKGRFGEVWRGRWRGGDVAVKIFSSREERSWFREAEIYQTVMLRHENILGFIAADNKDNGTWTQLWLVSDYHEHGSLFDYLNRYTVTIEGMIKLALSAASGLAHLHMEIVGTQGKPGIAHRDLKSKNILVKKNGMCAIADLGLAVRHDAVTDTIDIAPNQRVGTKRYMAPEVLDETINMKHFDSFKCADIYALGLVYWEIARRCNSGGVHEEYQLPYYDLVPSDPSIEEMRKVVCDQKLRPNIPNWWQSYEALRVMGKMMRECWYANGAARLTALRIKKTLSQLSVQEDVKI
Specifications

Specifications

AAH00254.1
activin A receptor, type IB
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACVR1B
GST
91
Wheat germ expression system
10 μg
ACTRIB, ACVRLK4, ALK4, SKR2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers