missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR3 Antibody (7E5), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | ACTR3 |
|---|---|
| Clone | 7E5 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043642
|
Novus Biologicals
NBP3-26525-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043065
|
Novus Biologicals
NBP3-26525-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACTR3 Monoclonal antibody specifically detects ACTR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| ACTR3 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| actin-related protein 3, ARP3 (actin-related protein 3, yeast) homolog, ARP3 actin-related protein 3 homolog (yeast), ARP3Actin-like protein 3 | |
| A synthesized peptide derived from Human ACTR3 [UniProt P61158] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7E5 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 10096 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title