Learn More
Novus Biologicals™ Activin RIIA Antibody (Ab-14E1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28281-1mg
 View more versions of this product
Product Code. 30044808
  • 920.00€ / 1mg
Description
Activin RIIA Monoclonal antibody specifically detects Activin RIIA in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Activin RIIA
Monoclonal
Unconjugated
P27037
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Ab-14E1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ACTRII, ACTRIIA, ACVR2ACTR-IIA, EC 2.7.11, EC 2.7.11.30
ACVR2A
1 mg
Cancer
92