missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin C/Inhibin beta C Antibody (CL6916) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76514PCP
This item is not returnable.
View return policy
Description
Activin C/Inhibin beta C Monoclonal antibody specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot
Specifications
| Activin C/Inhibin beta C | |
| Monoclonal | |
| PerCP | |
| inhibin beta C chain, IHBC, inhibin, beta C | |
| This antibody was developed using a recombinant protein derived from P55103, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cytokine Research | |
| 3626 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot | |
| CL6916 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction