missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin, Smooth Muscle Mouse, Clone: 1A4; same as asm-1, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 59-MSM1-P1
This item is not returnable.
View return policy
Specifications
| Actin, Smooth Muscle | |
| Cytoplasmic | |
| 1A4; same as asm-1 | |
| Unconjugated | |
| NP_001307784 | |
| Mouse | |
| 42kDa | |
| Protein A/G Purified | |
| RUO | |
| 59 | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| ACTA2 | |
| N-Terminal decapeptide of alpha smooth muscle isoform of actin, conjugated to KLH. | |
| Blood vessels in all tissues, smooth muscle or leiomyosarcoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction