missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Antibody (CLIP/1449) - C-terminus, Novus Biologicals™
Mouse Monoclonal Antibody
277.00€ - 530.00€
Specifications
| Antigen | Beta-endorphin |
|---|---|
| Clone | CLIP/1449 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204530
|
Novus Biologicals
NBP2-53161-20UG |
20 μg |
277.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18231430
|
Novus Biologicals
NBP2-53161-100UG |
100 μg |
530.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Beta-endorphin | |
| 0.2 mg/mL | |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Nutrient Sensing in the Brain | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 5443 | |
| Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
| Primary | |
| Store at 4C. |
| CLIP/1449 | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| POMC | |
| IgG1 | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title