missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL4 Rabbit anti-Human, Mouse, Clone: 3I9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 81196-1-RR-100UL
This item is not returnable.
View return policy
Description
ACSL4 Recombinant Monoclonal antibody specifically detects ACSL4 in Human, Mouse samples. It is validated for WB, ELISASpecifications
| ACSL4 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Rabbit | |
| 100 μL | |
| 2182 | |
| Store at -20°C | |
| Liquid |
| Western Blot, ELISA | |
| 3I9 | |
| Unconjugated | |
| BC038235 | |
| Protein A purification | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction