missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACP6 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149401
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Lysophosphatidic acid phosphatase type 6 (ACP6), also designated acid phosphatase-like protein 1 (ACPL1) or lysophosphatidic acid phosphatase (LPAP), is a 428 amino acid secreted protein that hydrolyzes lysophosphatidic acid to monoacylglycerol. ACP6 is highly expressed in kidney, heart, small intestine, muscle, liver, prostate, testis, ovary and exists as two isoforms as a result of alternative splicing events. The gene encoding ACP6 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene of human chromosome 1, which encodes lamin A. Stickler syndrome, Parkinsons, Gaucher disease, familial adenomatous polyposis and Usher syndrome are also associated with chromosome 1. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
Specifications
| ACP6 | |
| Polyclonal | |
| Unconjugated | |
| ACP6 | |
| 3.1.3.2; 5730559A09Rik; Acid phosphatase 6, lysophosphatidic; acid phosphatase like 1; acid phosphatase-like protein 1; Acp6; Acpl1; AU022842; Lpap; Lysophosphatidic acid phosphatase type 6; mPACPL1; PACPL1; UNQ205/PRO231 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 51205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NPH0 | |
| ACP6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ACP6, Glu33-Glu428. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction