Learn More
Acitretin, Mikromol™
Description
Acitretin
Specifications
Specifications
| Chemical Name or Material | Acitretin |
| CAS | 55079-83-9 |
| Analyte or Component Names | Acitretin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Acitretin, (all-E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid |
| InChI Formula | InChI=1S/C21H26O3/c1-14(8-7-9-15(2)12-21(22)23)10-11-19-16(3)13-20(24-6)18(5)17(19)4/h7-13H,1-6H3,(H,22,23)/b9-7+,11-10+,14-8+,15-12+ |
| SMILES | COc1cc(C)c(\C=C\C(=C\C=C\C(=C\C(=O)O)\C)\C)c(C)c1C |
| IUPAC Name | (2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid |
| Show More |