missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | ACF1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230773
|
Novus Biologicals
NBP3-38624-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231176
|
Novus Biologicals
NBP3-38624-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACF1 Polyclonal antibody specifically detects ACF1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| ACF1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ACF1hWALp1, ATP-dependent chromatin remodeling protein, ATP-dependent chromatin-remodeling protein, ATP-utilizing chromatin assembly and remodeling factor 1, bromodomain adjacent to zinc finger domain protein 1A, bromodomain adjacent to zinc finger domain, 1A, CHRAC subunit ACF1, hACF1FLJ14383, WALp1, WCRF180DKFZp586E0518, Williams syndrome transcription factor-related chromatin-remodeling factor 180 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1347-1556 of human ACF1 (NP_038476.2).,, Sequence:, VFVELLSPRRKRRGRKSANNTPENSPNFPNFRVIATKSSEQSRSVNIASKLSLQESESKRRCRKRQSPEPSPVTLGRRSSGRQGGVHELSAFEQLVVELVRHDDSWPFLKLVSKIQVPDYYDIIKKPIALNIIREKVNKCEYKLASEFIDDIELMFSNCFEYNPRNTSEAKAGTRLQAFFHIQAQKLGLHVTPSNVDQVSTPPAAKKSRI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 11177 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title