missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetamiprid

Product Code. p-7214476 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16584254 250 mg 250mg
16574254 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16584254 Supplier Thermo Scientific Chemicals Supplier No. 463932500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135410-20-7 | C10H11ClN4 | 222.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135410-20-7
Molecular Formula C10H11ClN4
Molecular Weight (g/mol) 222.68
InChI Key WCXDHFDTOYPNIE-UHFFFAOYSA-N
IUPAC Name N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
SMILES CN(CC1=CC=C(Cl)N=C1)C(C)=NC#N

Specifications

Melting Point 100.0°C to 102.0°C
Solubility Soluble in methanol, chloroform and dimethyl sulfoxide
Quantity 250 mg
Formula Weight 222.67
Chemical Name or Material Acetamiprid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acetamiprid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H361d-Suspected of damaging the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Acetamiprid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.